Anti-GOS-28 (GOLGI SNARE ) Rabbit Monoclonal Antibody

GTX61678
89364-126EA 556.26 CAD
89364-126
Anti-GOS-28 (GOLGI SNARE ) Rabbit Monoclonal Antibody
In eukaryotic cells, the Golgi apparatus receives newly synthesized proteins from the endoplasmic reticulum (ER) and delivers them after covalent modification to their destination in the cell. Although little is known about membrane-tubule-mediated transport, the molecular mechanism for vesicle-mediated transport is quite well understood, occurring through docking of SNAREs on the vesicle with those on the target membrane. . Transport of proteins along the exocytotic pathway is primarily achieved by vesicular intermediates. Two proteins, Golgi SNAREs of 27 kDa, GS27, and of 28 kDa, GS28 are important trafficking membrane proteins between the endoplasmic reticulum and the Golgi and between Golgi subcompartments . In vitro, GOS-28, A Golgi SNARE of 28 kD, is efficiently packaged into Golgi-derived vesicles, which are most likely COPI coated. Antibodies directed against GOS-28 block its ability to bind alpha-SNAP, partially inhibit transport from the cis to the medial cisternae, and do not inhibit budding of COP-coated vesicles, but do accumulate docked uncoated vesicles .

Type: Primary
Antigen: GOS-28 (GOLGI SNARE )
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR