Anti-GP IIB/IIIA Mouse Monoclonal Antibody [clone: 237]

Supplier: GeneTex
GTX20662
89360-864EA 997.99 CAD
89360-864
Anti-GP IIB/IIIA Mouse Monoclonal Antibody [clone: 237]
Antibodies
The integrins are a family of more than 23 heterodimeric transmembrane proteins that mediate cell-cell adhesions as well as cell-substratum adhesions and signal transduction processes. Integrins are heterodimers consisting of noncovalently associated alpha and beta subunits. CD61 non-covalently associates with CD41 (alpha Ilb integrin) and is expressed by megakaryocytes and platelets. The CD61/CD41 complex acts as a receptor for such adhesive ligands as fibronectin, fibrinogen and von Willebrand factor during platelet stimulation.

Type: Primary
Antigen: GP IIB/IIIA
Clonality: Monoclonal
Clone: [237]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR