Anti-GPR116 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX70551
89365-046EA 728.03 CAD
89365-046
Anti-GPR116 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal to Human G Protein-Coupled Receptor GPR116

For IHC-P: Use at a dilution of 2 - 4 ?g/ml.Optimal dilution to be determined by the researcher.

Type: Primary
Antigen: ADGRF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR