Anti-GPCR GPR88 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX78113
89366-506EA 664.1 CAD
89366-506
Anti-GPCR GPR88 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to G Protein-Coupled Receptor GPR88

Type: Primary
Antigen: GPR88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR