Anti-GPRC5C Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX108193
89355-358EA 881.1 CAD
89355-358
Anti-GPRC5C Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GPRC5C (G protein-coupled receptor, family C, group 5, member C)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GPRC5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR