Anti-GPI Mouse Monoclonal Antibody

GTX84418
89366-790EA 607.77 CAD
89366-790
Anti-GPI Mouse Monoclonal Antibody
Besides it's role as a glycolytic enzyme, mammalian GPI can function as a tumor-secreted cytokine and an angiogenic factor (AMF) that stimulates endothelial cell motility. GPI is also a neurotrophic factor (Neuroleukin) for spinal and sensory neurons.

Type: Primary
Antigen: GPI
Clonality: Monoclonal
Clone: "ERROR:#N/A"
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR