Anti-GH1 Rabbit Polyclonal Antibody

GTX74405
89365-448EA 521.86 CAD
89365-448
Anti-GH1 Rabbit Polyclonal Antibody
Recognizes human growth hormone (GH), which acts to control body growth and metabolic activity. Growth hormone is a small peptide hormone released by the anterior pituitary under the control the hypothalamus. It recognizes GH producing cells of the pituitary and neoplasms arising from these cells.

Type: Primary
Antigen: Growth Hormone
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR