Anti-GRPR Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100015
89348-126EA 881.1 CAD
89348-126
Anti-GRPR Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to GRPR (gastrin-releasing peptide receptor)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: GRPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR