Anti-GSK3A Rabbit Polyclonal Antibody

GTX29765
89367-650EA 701.22 CAD
89367-650
Anti-GSK3A Rabbit Polyclonal Antibody
Glycogen synthase kinase-3 (GSK 3) is a proline-directed serine-threonine kinase that was initially identified as a phosphorylating and inactivating glycogen synthase. Two isoforms, alpha (GSK 3A) and beta, show a high degree of amino acid homology. GSK 3B is involved in energy metabolism, neuronal cell development, and body pattern formation.

Type: Primary
Antigen: GSK3 alpha (Phospho SER21)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR