Anti-GSK3B Rabbit Monoclonal Antibody

GTX62011
89364-270EA 556.26 CAD
89364-270
Anti-GSK3B Rabbit Monoclonal Antibody
Glycogen synthase kinase 3 beta (GSK3 beta) is a Serine/Threonine protein kinase involved in the insulin and wingless pathways . Originally identified as a regulator of glycogen synthase, it has been shown to regulate a diverse array of cellular functions. GSK3 beta activity is down regulated by phosphorylation on Serine 9 by PKB, or activated by phosphorylation on Tyrosine 216. Once activated, the protein can phosphorylate p53, c-jun, heat shock factor-1 and cyclin D1 . It is also known to phosphorylate Tau in Allzheimers disease. Additionally, increased GSK3 beta protein levels are found in Alzheimers disease brains.

Type: Primary
Antigen: GSK3 beta (Phospho SER9)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR