Anti-GSK3B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX48623
89352-894EA 441.11 CAD
89352-894
Anti-GSK3B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GSK3 beta

Western Blot 0.5 ug/ml,

Type: Primary
Antigen: GSK3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR