Anti-GSPT1 Rabbit Polyclonal Antibody

GTX111060
89356-538EA 476 CAD
89356-538
Anti-GSPT1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: GSPT1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR