Anti-GST Mouse Monoclonal Antibody

Supplier: GeneTex
GTX18183
89360-424EA 998.26 CAD
89360-424
Anti-GST Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: GST
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR