Anti-HA TAG Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX115044-01
89358-226EA 640.44 CAD
89358-226
Anti-HA TAG Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HA TAG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR