Anti-HDC Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX112435
89357-072EA 673.08 CAD
89357-072
Anti-HDC Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to HDC (histidine decarboxylase)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HDC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR