Anti-HelicobacterR Pylori Mouse Monoclonal Antibody

GTX28081
89352-340EA 466.29 CAD
89352-340
Anti-HelicobacterR Pylori Mouse Monoclonal Antibody
The bacterium Helicobater Pylori is strongly associated with inflammation of the stomach and is also implicated in the development of gastric malignancy.

Type: Primary
Antigen: HelicobacterR Pylori
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bacteria
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR