Anti-HBA1 Mouse Monoclonal Antibody [clone: B842M]

Supplier: GeneTex
GTX42177
89363-498EA 856.63 CAD
89363-498
Anti-HBA1 Mouse Monoclonal Antibody [clone: B842M]
Antibodies
Mouse Monoclonal antibody [B842M] to Hemoglobin A1c (HbA1c)

Recommended antibody pair for sandwich ELISA:<br />GTX42115 for capture antibody , and GTX42177 for detection antibody

Type: Primary
Antigen: HBA1
Clonality: Monoclonal
Clone: B842M
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR