Anti-HBA1 Rabbit Monoclonal Antibody

GTX62239
89364-364EA 585.93 CAD
89364-364
Anti-HBA1 Rabbit Monoclonal Antibody
Hemoglobin alpha chain belongs to the globin family and is involved in oxygen transport from the lung to the various peripheral tissues. It is a heterotetramer of two alpha chains and two beta chains in adult hemoglobin A (HbA); two alpha chains and two delta chains in adult hemoglobin A2 (HbA2). Hemoglobin alpha chain is expressed in red blood cells and defects in HBA1/HBA2 can lead to alpha thalassemia, the most common of monogenic diseases .

Type: Primary
Antigen: Hemoglobin alpha Chain
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR