Anti-Hepatitis B Mouse Monoclonal Antibody

GTX75528
89367-888EA 593.84 CAD
89367-888
Anti-Hepatitis B Mouse Monoclonal Antibody
Hepatitis B virus is a major causative agent of acute and chronic liver disease in humans. Hepatitis B surface antigen (HBsAg) is a protein component of the viral envelope, which is expressed predominantly in the cytoplasm of infected hepatocytes.

Type: Primary
Antigen: Hepatitis B
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR