Anti-Hepatitis C Core Antigen Mouse Monoclonal Antibody

Supplier: GeneTex
GTX22584
89351-812EA 992.08 CAD
89351-812
Anti-Hepatitis C Core Antigen Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Hepatitis C Core Antigen
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR