Anti-Hepatitis C NS3 Mouse Monoclonal Antibody

Supplier: GeneTex

GTX78171
89366-526EA 789.92 CAD
89366-526
Anti-Hepatitis C NS3 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: Hepatitis C NS3
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR