Anti-Hepatitis C NS5 Goat Polyclonal Antibody

Supplier: GeneTex
GTX40773
89363-302EA 599.3 CAD
89363-302
Anti-Hepatitis C NS5 Goat Polyclonal Antibody
Antibodies
NS5 may play a role in the viral RNA replication of Hepatitis C Virus.

Type: Primary
Antigen: Hepatitis C NS5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR