Anti-Hepatitis G Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX70318
89364-904EA 580.76 CAD
89364-904
Anti-Hepatitis G Rabbit Polyclonal Antibody
Antibodies
Hepatitis G Virus (also designated GBV-C/HBV).

Type: Primary
Antigen: Hepatitis G
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR