Anti-HAS Mouse Monoclonal Antibody [clone: OCH1E5]

Supplier: GeneTex
GTX73779
89353-274EA 1290.01 CAD
89353-274
Anti-HAS Mouse Monoclonal Antibody [clone: OCH1E5]
Antibodies
Hepatoblastoma is the most common primary tumor of the liver in children. The use of specific hepatocyte markers and also of alpha Fetoprotein or carcinoembryonic antigen are useful for the identification of normal and malignant fetal hepatocytes. This antibody recognizes an uncharacterized antigen present in both adults and fetal normal hepatocytes to produce a distinct granular cytoplasmic staining. This antibody stains majority of hepatocellular carcinomas.

Type: Primary
Antigen: Hepatocyte SSpecific Antigen
Clonality: Monoclonal
Clone: [OCH1E5]
Conjugation: No
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR