Anti-HER3 Rabbit Monoclonal Antibody

GTX62097
89364-318EA 556.26 CAD
89364-318
Anti-HER3 Rabbit Monoclonal Antibody
HER3/ErbB3 is a member of the ErbB/EGF receptor protein tyrosine kinase family . Tyrosine phosphorylation of HER3/ErbB3 depends on its association with other ErbB tyrosine kinases . Phosphorylation sites in the carboxy-terminal tail mediate downstream signaling pathways. HER3/ErbB3 is highly expressed in many cancer cells , and it may work together with other Erb members in tumorigenesis and proliferation . HER/ErbB3 is phosphorylated by EGFR or ErbB2 through dimeric interactions with HER/ErbB3, on Tyrosine 1289, a docking site for phosphatidylinositol 3-kinase .

Type: Primary
Antigen: HER3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR