Anti-HES1 Rabbit Monoclonal Antibody

Supplier: Genetex
GTX62458
89364-456EA 615.46 CAD
89364-456
Anti-HES1 Rabbit Monoclonal Antibody
Rabbit monoclonal antibody [EPR4226] to HES1

WB: 1/1000. Flow Cyt: 1/50. ICC/IF: 1/100. IHC-P: 1/200. Perform heat mediated antigen retrieval before commencing with IHC staining protocol. Is unsuitable for IP.

Type: Primary
Antigen: HES1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR