Anti-HIF1A Mouse Monoclonal Antibody

GTX21817
89361-084EA 518.73 CAD
89361-084
Anti-HIF1A Mouse Monoclonal Antibody
HIF-1 is a key component of a widely operative transcriptional response activated by hypoxia, cobaltous ions and iron chelation. HIF-1 activates transcription of hypoxia-inducible genes.

Type: Primary
Antigen: HIF1A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR