Anti-HRH4 Rabbit Polyclonal Antibody

GTX71877
89365-340EA 470.87 CAD
89365-340
Anti-HRH4 Rabbit Polyclonal Antibody
Histamine Receptor H4 binds histamine and several psychoactive compounds such as amitriptyline, chlorpromazine, cyproheptadine, and mianserin. The Histamine H4 receptor has been reported in human blood peripheral leukocytes, bone marrow, colon, liver, lung, smalll intestine, spleen, testis, thymus, tonsil, and trachea. Contrary to the mouse H4 receptor, the human gene was not detected in brain.

Type: Primary
Antigen: HRH4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR