Anti-HIV Goat Polyclonal Antibody

Supplier: GeneTex
GTX40715
89363-276EA 956.96 CAD
89363-276
Anti-HIV Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HIV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR