Anti-HLA Mouse Monoclonal Antibody [clone: D1]

GTX74621
89367-802EA 627.65 CAD
89367-802
Anti-HLA Mouse Monoclonal Antibody [clone: D1]
Mouse monoclonal antibody [D1] to MHC Class I H-2Dd

Type: Primary
Antigen: HLA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR