Anti-HLA Mouse Monoclonal Antibody [clone: K3]

Supplier: GeneTex
GTX74622
89367-804EA 1138.22 CAD
89367-804
Anti-HLA Mouse Monoclonal Antibody [clone: K3]
Antibodies
Mouse monoclonal antibody [K3] to MHC Class I H-2Kd

Type: Primary
Antigen: HLA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR