Anti-HLA-DR + HLA-DP Mouse Monoclonal Antibody

Supplier: GeneTex
GTX18232
89360-436EA 673.68 CAD
89360-436
Anti-HLA-DR + HLA-DP Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: HLA-DR + HLA-DP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR