Anti-HLA-DR Mouse Monoclonal Antibody [clone: MEM-12]

Supplier: GeneTex

GTX78432
89366-644EA 480.17 CAD
89366-644
Anti-HLA-DR Mouse Monoclonal Antibody [clone: MEM-12]
Antibodies
Monoclonal Antibody to HLA-DR (Human) Purified Antibody

FACS: Positive control: B-lymphocytes, Monocytes, DAUDI cell line, RAJI cell lineIP: Positive control: B-lymphocytes, Monocytes, DAUDI cell line, RAJI cell lineWB: Positive control: B-lymphocytes, Monocytes, DAUDI cell line, RAJI cell lineApplication note: non-boiled, non-reducing conditions

Type: Primary
Antigen: HLA-DR
Clonality: Monoclonal
Clone: MEM-12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR