Anti-HLA-DR beta Mouse Monoclonal Antibody

Supplier: GeneTex

GTX40182
89363-124EA 640.88 CAD
89363-124
Anti-HLA-DR beta Mouse Monoclonal Antibody
Antibodies
Human Leukocyte Antigens are highly polymorphic proteins that are involved in the presentation of antigens to the T-cell receptor. There are two classes of HLA antigens, class I (HLA-A, HLA-B and HLA-C) and class II (HLA-D). TDR 31.1 can be used for the identification of class II HLA molecules. Not cytotoxic with rabbit complement.

Type: Primary
Antigen: HLA-DR beta
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR