Anti-HLTF Rabbit Polyclonal Antibody

GTX17984
89360-364EA 591.15 CAD
89360-364
Anti-HLTF Rabbit Polyclonal Antibody
The HLTF (Helicase-like Transcription Factor; SMARCA 3) gene is a member of the family of genes that remove proteins that coil around DNA. HLTF proteins aid the stabilization and regulation of the production of proteins in the cell.Inactivation of the HTLF gene may contribute to the transformation of normal colon cells into cancer cells.

Type: Primary
Antigen: HLTF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR