Anti-HMGCS2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX47146
89363-622EA 787.36 CAD
89363-622
Anti-HMGCS2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to HMGCS2 (N-terminal)

Recommended Dilutions: For Western Blot: Use at 5.0 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: HMGCS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR