Anti-HMW Kininogen Heavy Chain Mouse Monoclonal Antibody [clone: 1.B.708]

GTX14512
89360-024EA 613.94 CAD
89360-024
Anti-HMW Kininogen Heavy Chain Mouse Monoclonal Antibody [clone: 1.B.708]
Kininogens are a group of inactive plasma proteins that, when proteolytically cleaved by Kallikrein, give rise to Kinins (Bradykinin and Kallidin). HMW Kininogen Heavy Chain is a coagulation protein involved in the early stages of intrinsic pathway activation. HMW Kininogen Heavy Chain deficiency can cause a marked prolongation of the PTT, but it does not cause bleeding. The same is true for factor XII deficiency and prekallikrein deficiency.

Type: Primary
Antigen: HMW Kininogen Heavy Chain
Clonality: Monoclonal
Clone: 1.B.708
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR