Anti-HRP Mouse Polyclonal Antibody

Supplier: Genetex
GTX22110
89361-164EA 1551.39 CAD
89361-164
Anti-HRP Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HRP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR