Anti-HP1 gamma Rabbit Polyclonal Antibody

GTX106434
89354-690EA 472.59 CAD
89354-690
Anti-HP1 gamma Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to HP1 gamma (chromobox homolog 3 (HP1 gamma homolog, Drosophila))

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HP1 gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR