Anti-HRH3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX13015
89359-552EA 701.31 CAD
89359-552
Anti-HRH3 Rabbit Polyclonal Antibody
Antibodies
Histamine is a ubiquitous messenger molecule released from mast cells, enterochromaffin-like cells, and neurons. Histamine receptor H3 belongs to the family 1 of G protein-coupled receptors. It is an integral membrane protein and can regulate neurotransmitter release. This receptor can also increase voltage-dependent calcium current in smooth muscles and innervates the blood vessels and the heart in cardiovascular system.

IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: HRH3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR