Anti-HSP70 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX22412
89351-806EA 548.82 CAD
89351-806
Anti-HSP70 Rabbit Polyclonal Antibody
Antibodies
This antibody may be diluted to a titer of 1:75-1:250 in an ABC method. We suggest an incubation period of 30 minutes at room temperature.

Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR