Anti-HSP90B Rabbit Polyclonal Antibody

GTX77733
89366-272EA 516.95 CAD
89366-272
Anti-HSP90B Rabbit Polyclonal Antibody
Rabbit Polyclonal Antibody to HSP 90 beta

WB: Suggested dilution at 1 ?g/ml in 5% skim milk / PBS buffer. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: HSP90B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR