Anti-HSV1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX29533
89352-634EA 978.11 CAD
89352-634
Anti-HSV1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to HSV1

This antibody may be diluted to a titer of 1:75-1:200 in an ABC method. We suggest an incubation period of 30-60 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation and dilutions should be determined by the user.

Type: Primary
Antigen: HSV1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR