Anti-HSV2 Mouse Monoclonal Antibody [clone: AP1 (B019)]

Supplier: GeneTex
GTX76112
89365-992EA 877.04 CAD
89365-992
Anti-HSV2 Mouse Monoclonal Antibody [clone: AP1 (B019)]
Antibodies
Mouse monoclonal antibody [AP1 (B019)] to HSV2

Recommended Starting Dilutions: Immunohistology - Frozen: Max Dilution 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: HSV2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR