Anti-ALB Mouse Monoclonal Antibody [clone: 1C8]

Supplier: GeneTex

GTX10243
89350-736EA 440.1 CAD
89350-736
Anti-ALB Mouse Monoclonal Antibody [clone: 1C8]
Antibodies
Mouse monoclonal antibody [1C8] to Albumin

ELISA: Use at an assay dependent dilution (capture, detection), produces sensitive assay system with rapid kinetics (10-15 minutes) for albumin in human urine. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: ALB
Clonality: Monoclonal
Clone: 1C8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR