Anti-IgE Sheep Polyclonal Antibody

Supplier: GeneTex
GTX77501
89353-750EA 319.71 CAD
89353-750
Anti-IgE Sheep Polyclonal Antibody
Antibodies
Sheep anti-Human IgE polyclonal antibody

Works in most applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Secondary
Antigen: IgE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR