Anti-IVNS1ABP Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX77835
89366-338EA 754.71 CAD
89366-338
Anti-IVNS1ABP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Antibody to Influenza Virus NS1A Binding Protein (IVNS1ABP)

IHC-P: Use at 4-8?g/ml. Use human stomach as positive control. WB: Use at 2.0?g/ml in 5% skim milk/PBS buffer. Use HepG2 as positive control. Not yet tested in other applications. Optimal dilutions should be determined by end-user.

Type: Primary
Antigen: IVNS1ABP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR