Anti-ING4 Rabbit Monoclonal Antibody [clone: EP3804]

Supplier: Genetex
GTX62655
89364-550EA 961.95 CAD
89364-550
Anti-ING4 Rabbit Monoclonal Antibody [clone: EP3804]
Antibodies
Rabbit monoclonal antibody [EP3804] to ING4

Recommended Starting Dilutions: For WB: Use at a dilution of 1:1,000 - 10,000 For IP: Use at a dilution of 1:10 - 100 Optimal working dilution for a specific application should be determined by the investigator.

Type: Primary
Antigen: ING4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR