Anti-INS Mouse Monoclonal Antibody [clone: D4B8]

Supplier: GeneTex
GTX28303
89352-426EA 1052.6 CAD
89352-426
Anti-INS Mouse Monoclonal Antibody [clone: D4B8]
Antibodies
Mouse monoclonal [D4B8] to Insulin

This antibody recognises a different epitope GTX28302 (7F8)Immunohistochemistry on frozen sections.

Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: D4B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Pig, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR