Anti-MAD2L2 Rabbit Polyclonal Antibody

Supplier: GeneTex

GTX23630
89361-362EA 638.13 CAD
89361-362
Anti-MAD2L2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to MAD2L2

Recommended Starting Dilutions: For ELISA: Use at a dilution of 1:7,500 - 1:30,000. For WB: Use at a dilution of 1:500 - 1:2,000. For IHC-P: Use at a dilution of 1:500 - 1:2,000. Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MAD2L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR